Product Details

SNP ID
rs2645673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76895076 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGCTACTAGCCAGAGAGGATTTT[A/G]ATTGTGCTTCATCAATTCCTCGAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100421494 PubMed Links

Gene Details

Gene
LOC100421494
Gene Name
microtubule affinity regulating kinase 2 pseudogene
There are no transcripts associated with this gene.

Gene
SOWAHB
Gene Name
sosondowah ankyrin repeat domain family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029870.1 2774 UTR 3 NP_001025041.1

View Full Product Details