Product Details

SNP ID
rs2723613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:133836801 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGTTCTACCAGGGCAGGGACCA[A/G]GTGGGTCTTATTTATTGCTGTATTC
Phenotype
MIM: 609805
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATA19 PubMed Links

Gene Details

Gene
SPATA19
Gene Name
spermatogenesis associated 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291992.1 Intron NP_001278921.1
NM_174927.2 Intron NP_777587.1

View Full Product Details