Product Details

SNP ID
rs2732705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:46274563 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGTACAGGAATCATTGATAAGATA[G/T]GGATGGATTATATGTAACCGTGGCT
Phenotype
MIM: 616555
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARL17B PubMed Links

Gene Details

Gene
ARL17B
Gene Name
ADP ribosylation factor like GTPase 17B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039083.3 1732 Intron NP_001034172.3
NM_001103154.1 1732 Intron NP_001096624.1
XM_005256918.4 1732 UTR 3 XP_005256975.1
XM_011524160.2 1732 Intron XP_011522462.1
XM_011524161.2 1732 Intron XP_011522463.1
XM_011524162.2 1732 UTR 3 XP_011522464.1
XM_011524163.2 1732 Intron XP_011522465.1
XM_011524164.2 1732 Intron XP_011522466.1
XM_011524165.2 1732 Intron XP_011522467.1
XM_011524166.2 1732 Intron XP_011522468.1
XM_011524167.2 1732 Intron XP_011522469.1
Gene
LRRC37A
Gene Name
leucine rich repeat containing 37A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014834.4 1732 Intron NP_055649.4
XM_005257879.4 1732 Intron XP_005257936.1
XM_005257880.4 1732 Intron XP_005257937.1
XM_005257882.4 1732 Intron XP_005257939.1
XM_005257883.4 1732 Intron XP_005257940.1
XM_005257884.4 1732 Intron XP_005257941.1
XM_006722210.3 1732 Intron XP_006722273.1
XM_006722211.3 1732 Intron XP_006722274.1
XM_011525539.2 1732 Intron XP_011523841.1
XM_017025472.1 1732 Intron XP_016880961.1
XM_017025473.1 1732 Intron XP_016880962.1

View Full Product Details