Product Details

SNP ID
rs2161630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69572558 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCAGAGAAAAGGCACCATTTGTT[A/G]TACATTCTATAATGAAAACTATTCA
Phenotype
MIM: 604708
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1538 PubMed Links

Gene Details

Gene
MIR1538
Gene Name
microRNA 1538
There are no transcripts associated with this gene.

Gene
NFAT5
Gene Name
nuclear factor of activated T-cells 5, tonicity-responsive
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113178.2 Intron NP_001106649.1
NM_006599.3 Intron NP_006590.1
NM_138713.3 Intron NP_619727.2
NM_138714.3 Intron NP_619728.2
NM_173214.2 Intron NP_775321.1
NM_173215.2 Intron NP_775322.1
XM_006721125.3 Intron XP_006721188.1
XM_011522817.2 Intron XP_011521119.1
XM_011522818.2 Intron XP_011521120.1
XM_011522819.2 Intron XP_011521121.1
XM_011522820.2 Intron XP_011521122.1
XM_011522822.2 Intron XP_011521124.1
XM_017022870.1 Intron XP_016878359.1
XM_017022871.1 Intron XP_016878360.1
XM_017022872.1 Intron XP_016878361.1
XM_017022873.1 Intron XP_016878362.1

View Full Product Details