Product Details

SNP ID
rs2182673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102602230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTTTATAAAGTAGTATCAACTT[A/T]ATTTTGTTTCCATTTTTATTTTGGT
Phenotype
MIM: 190470
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TPP2 PubMed Links

Gene Details

Gene
TPP2
Gene Name
tripeptidyl peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005254070.1 Intron XP_005254127.1
XM_005254071.1 Intron XP_005254128.1
XM_005254072.1 Intron XP_005254129.1

View Full Product Details