Product Details
- SNP ID
-
rs2106579
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31721767 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTCCACATCATCCCTGAATTCTTC[G/C]TTCAGTGTTCAACCTCTCTGCTATT
- Phenotype
-
- Polymorphism
- G/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF720
PubMed Links
Gene Details
- Gene
- ZNF720
- Gene Name
- zinc finger protein 720
View Full Product Details