Product Details

SNP ID
rs2857657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34256113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTGGGATTTAATGAGCTCTTT[C/G]TCTTCTCCTGCCTGCCTTTTGCTTT
Phenotype
MIM: 158105
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCL2 PubMed Links

Gene Details

Gene
CCL2
Gene Name
C-C motif chemokine ligand 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002982.3 Intron NP_002973.1

View Full Product Details