Product Details

SNP ID
rs2074592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5587166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTTACTTTGCTTTTAAAAAGAT[A/C]CCCCAAGTTCGAGGGAACCCTGGCT
Phenotype
MIM: 608066
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SAFB2 PubMed Links

Gene Details

Gene
SAFB2
Gene Name
scaffold attachment factor B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014649.2 2501 UTR 3 NP_055464.1
XM_011528449.2 2501 UTR 3 XP_011526751.1
XM_011528450.2 2501 UTR 3 XP_011526752.1
XM_017027520.1 2501 UTR 3 XP_016883009.1
XM_017027521.1 2501 UTR 3 XP_016883010.1
XM_017027522.1 2501 UTR 3 XP_016883011.1

View Full Product Details