Product Details

SNP ID
rs2076045
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32416556 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAGAAATGCTGTCCATTTTTAGC[C/T]TACCCGATGTATATTTTGAAAATAC
Phenotype
MIM: 614109 MIM: 613901
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
BPIFC PubMed Links

Gene Details

Gene
BPIFC
Gene Name
BPI fold containing family C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174932.2 Intron NP_777592.1
XM_011530088.2 Intron XP_011528390.1
XM_011530089.1 Intron XP_011528391.1
XM_011530090.1 Intron XP_011528392.1
XM_011530091.2 Intron XP_011528393.1
XM_017028740.1 Intron XP_016884229.1
Gene
RTCB
Gene Name
RNA 2',3'-cyclic phosphate and 5'-OH ligase
There are no transcripts associated with this gene.

View Full Product Details