Product Details

SNP ID
rs2964575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151774541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTTTTTCTTTATTTTTGTCAG[A/T]CTGATAAAACTGCCCACTTGGTCTT
Phenotype
MIM: 608431
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
G3BP1 PubMed Links

Gene Details

Gene
G3BP1
Gene Name
G3BP stress granule assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005754.2 Intron NP_005745.1
NM_198395.1 Intron NP_938405.1
XM_006714749.3 Intron XP_006714812.1
XM_006714750.3 Intron XP_006714813.1
XM_017008922.1 Intron XP_016864411.1
XM_017008923.1 Intron XP_016864412.1
Gene
LOC100652758
Gene Name
uncharacterized LOC100652758
There are no transcripts associated with this gene.

View Full Product Details