Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001243746.1 | 1975 | UTR 3 | NP_001230675.1 | ||
NM_017565.3 | 1975 | UTR 3 | NP_060035.2 | ||
XM_006721959.2 | 1975 | UTR 3 | XP_006722022.1 | ||
XM_011524918.2 | 1975 | Intron | XP_011523220.1 | ||
XM_017024781.1 | 1975 | Intron | XP_016880270.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001276289.1 | 1975 | Intron | NP_001263218.1 | ||
NM_001276290.1 | 1975 | Intron | NP_001263219.1 | ||
NM_001278433.1 | 1975 | Intron | NP_001265362.1 | ||
NM_002734.4 | 1975 | Intron | NP_002725.1 | ||
NM_212471.2 | 1975 | Intron | NP_997636.1 | ||
NM_212472.2 | 1975 | Intron | NP_997637.1 | ||
XM_011524983.2 | 1975 | Intron | XP_011523285.1 | ||
XM_011524984.2 | 1975 | Intron | XP_011523286.1 | ||
XM_011524985.2 | 1975 | Intron | XP_011523287.1 |