Product Details

SNP ID
rs2294053
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:125530496 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAACAGAGAAGAACCTCATTTGGTC[A/G]AGTTTTAGATAGTATGTGTCACTCT
Phenotype
MIM: 606739
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSBPL11 PubMed Links

Gene Details

Gene
OSBPL11
Gene Name
oxysterol binding protein like 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022776.4 3000 UTR 3 NP_073613.2

View Full Product Details