Product Details

SNP ID
rs2294372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:25354567 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCATGCAAGTCTGCAGCTGAGCAA[C/T]GGCTTGGGGCCCACCCCCCTGGACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100652901 PubMed Links

Gene Details

Gene
LOC100652901
Gene Name
uncharacterized LOC100652901
There are no transcripts associated with this gene.

Gene
LRP5L
Gene Name
LDL receptor related protein 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135772.1 Intron NP_001129244.1
NM_182492.2 Intron NP_872298.1
XM_006724372.3 Intron XP_006724435.1
XM_017029083.1 Intron XP_016884572.1
XM_017029084.1 Intron XP_016884573.1
XM_017029085.1 Intron XP_016884574.1
XM_017029086.1 Intron XP_016884575.1
XM_017029087.1 Intron XP_016884576.1
XM_017029088.1 Intron XP_016884577.1

View Full Product Details