Product Details

SNP ID
rs2446716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:59252014 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGAATACTCATCAGCATTTGGT[C/G]ATAGACTGAACATAAGAGATACAGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM13C PubMed Links

Gene Details

Gene
FAM13C
Gene Name
family with sequence similarity 13 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001971.2 Intron NP_001001971.1
NM_001143773.1 Intron NP_001137245.1
NM_001166698.1 Intron NP_001160170.1
NM_198215.3 Intron NP_937858.2
XM_005269618.4 Intron XP_005269675.2
XM_005269619.4 Intron XP_005269676.2
XM_006717702.3 Intron XP_006717765.1
XM_006717703.3 Intron XP_006717766.1
XM_011539490.2 Intron XP_011537792.1
XM_011539491.2 Intron XP_011537793.1
XM_017015885.1 Intron XP_016871374.1
XM_017015886.1 Intron XP_016871375.1
XM_017015887.1 Intron XP_016871376.1
XM_017015888.1 Intron XP_016871377.1
XM_017015889.1 Intron XP_016871378.1
XM_017015890.1 Intron XP_016871379.1
XM_017015891.1 Intron XP_016871380.1
XM_017015892.1 Intron XP_016871381.1
XM_017015893.1 Intron XP_016871382.1
Gene
PHYHIPL
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein like
There are no transcripts associated with this gene.

View Full Product Details