Product Details

SNP ID
rs2735392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31181506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTAAGGCGAGCAGTTGCATGAT[G/C]TCTGTCATTTGGGGTCCAAGGGCTC
Phenotype
MIM: 137070
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
FUS PubMed Links

Gene Details

Gene
FUS
Gene Name
FUS RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170634.1 Intron NP_001164105.1
NM_001170937.1 Intron NP_001164408.1
NM_004960.3 Intron NP_004951.1
XM_005255233.4 Intron XP_005255290.1
XM_011545781.1 Intron XP_011544083.1
XM_011545782.2 Intron XP_011544084.1

View Full Product Details