Product Details

SNP ID
rs73837443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51388885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGCTCCACCCAGTCAGTGACT[A/C]TCCCTGCTCTCTCCTCCAGACAAGC
Phenotype
MIM: 603123 MIM: 601916 MIM: 612602
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCAF1 PubMed Links

Gene Details

Gene
DCAF1
Gene Name
DDB1 and CUL4 associated factor 1
There are no transcripts associated with this gene.

Gene
DOCK3
Gene Name
dedicator of cytokinesis 3
There are no transcripts associated with this gene.

Gene
MANF
Gene Name
mesencephalic astrocyte derived neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006010.5 Intron NP_006001.4
Gene
RBM15B
Gene Name
RNA binding motif protein 15B
There are no transcripts associated with this gene.

View Full Product Details