Product Details

SNP ID
rs61739570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:25598337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACATTGCAAAATCAACATCATC[A/T]TCTTCTTCATCAGACTCTGCAAAAT
Phenotype
MIM: 180220 MIM: 126431
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RARB PubMed Links

Gene Details

Gene
RARB
Gene Name
retinoic acid receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000965.4 5490 Intron NP_000956.2
NM_001290216.2 5490 Intron NP_001277145.1
NM_001290217.1 5490 Intron NP_001277146.1
NM_001290266.1 5490 Intron NP_001277195.1
NM_001290276.1 5490 Intron NP_001277205.1
NM_001290277.1 5490 Intron NP_001277206.1
NM_001290300.1 5490 Intron NP_001277229.1
NM_016152.3 5490 Intron NP_057236.1
Gene
TOP2B
Gene Name
topoisomerase (DNA) II beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001068.3 5490 Missense Mutation GAA,GAT E1612D NP_001059.2
XM_005265427.3 5490 Missense Mutation GAA,GAT E1617D XP_005265484.1
XM_011534057.2 5490 Missense Mutation GAA,GAT E1580D XP_011532359.1

View Full Product Details