Product Details

SNP ID
rs34512715
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:26794649 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTGCCACTGGGGTACGCTCCAA[C/T]GGGCTGGTCAGTGTTGGCTTCCTCA
Phenotype
MIM: 610274
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIGV PubMed Links

Gene Details

Gene
PIGV
Gene Name
phosphatidylinositol glycan anchor biosynthesis class V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202554.1 1304 Silent Mutation AAC,AAT N205N NP_001189483.1
NM_017837.3 1304 Silent Mutation AAC,AAT N205N NP_060307.2

View Full Product Details