Product Details

SNP ID
rs112303272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:106325169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATACTTAAAATGAATTCATACA[C/T]TGATGGAGAAAAAATAATGTTTACC
Phenotype
MIM: 603605 MIM: 616899
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIMP1 PubMed Links

Gene Details

Gene
AIMP1
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142415.1 Intron NP_001135887.1
NM_001142416.1 Intron NP_001135888.1
NM_004757.3 Intron NP_004748.2
XM_017008835.1 Intron XP_016864324.1
XM_017008836.1 Intron XP_016864325.1
Gene
TBCK
Gene Name
TBC1 domain containing kinase
There are no transcripts associated with this gene.

View Full Product Details