Product Details

SNP ID
rs61736884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:29123498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGCCACCAAAACCCTGGCATC[C/T]GCATAGGCCTTGCTGTCCACTAAAG
Phenotype
MIM: 602712
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APBA2 PubMed Links

Gene Details

Gene
APBA2
Gene Name
amyloid beta precursor protein binding family A member 2
There are no transcripts associated with this gene.

Gene
FAM189A1
Gene Name
family with sequence similarity 189 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015307.1 1618 Silent Mutation GCA,GCG A487A NP_056122.1
XM_011521407.2 1618 Silent Mutation GCA,GCG A524A XP_011519709.1

View Full Product Details