Product Details

SNP ID
rs34643721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75717926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCCTTGTTTATGCCGTGGGAGC[A/G]CAGGCTGCAGACACTTGCTGTCCTA
Phenotype
MIM: 610270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOGAT2 PubMed Links

Gene Details

Gene
MOGAT2
Gene Name
monoacylglycerol O-acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025098.3 108 Missense Mutation CAC,CGC H13R NP_079374.2
XM_011545267.1 108 Missense Mutation CAC,CGC H13R XP_011543569.1

View Full Product Details