Product Details
- SNP ID
-
rs62636632
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44722944 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGTGCCAGGAAGCATCCCTTTTC[C/T]GGAAAGTCCTTTTACTTGGATCTGC
- Phenotype
-
MIM: 611661
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DBF4B
PubMed Links
Gene Details
- Gene
- DBF4B
- Gene Name
- DBF4 zinc finger B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025104.3 |
333 |
Silent Mutation |
TCC,TCT |
S49S |
NP_079380.2 |
NM_145663.2 |
333 |
Silent Mutation |
TCC,TCT |
S49S |
NP_663696.1 |
XM_006722098.2 |
333 |
Silent Mutation |
TCC,TCT |
S49S |
XP_006722161.1 |
XM_006722102.3 |
333 |
UTR 5 |
|
|
XP_006722165.1 |
XM_011525281.2 |
333 |
UTR 5 |
|
|
XP_011523583.1 |
XM_017025145.1 |
333 |
Silent Mutation |
TCC,TCT |
S49S |
XP_016880634.1 |
XM_017025146.1 |
333 |
Silent Mutation |
TCC,TCT |
S33S |
XP_016880635.1 |
XM_017025147.1 |
333 |
Silent Mutation |
TCC,TCT |
S28S |
XP_016880636.1 |
XM_017025148.1 |
333 |
Silent Mutation |
TCC,TCT |
S49S |
XP_016880637.1 |
XM_017025149.1 |
333 |
Silent Mutation |
TCC,TCT |
S49S |
XP_016880638.1 |
XM_017025150.1 |
333 |
Silent Mutation |
TCC,TCT |
S49S |
XP_016880639.1 |
XM_017025151.1 |
333 |
Silent Mutation |
TCC,TCT |
S49S |
XP_016880640.1 |
XM_017025152.1 |
333 |
UTR 5 |
|
|
XP_016880641.1 |
XM_017025153.1 |
333 |
UTR 5 |
|
|
XP_016880642.1 |
XM_017025154.1 |
333 |
UTR 5 |
|
|
XP_016880643.1 |
XM_017025155.1 |
333 |
UTR 5 |
|
|
XP_016880644.1 |
XM_017025156.1 |
333 |
Silent Mutation |
TCC,TCT |
S49S |
XP_016880645.1 |
View Full Product Details