Product Details

SNP ID
rs74693271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49703040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAGATAAAGGTGAGGTCCGGT[A/G]GGCTTCTGGGTATCTGCCATTGGTC
Phenotype
MIM: 610790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM117A PubMed Links

Gene Details

Gene
FAM117A
Gene Name
family with sequence similarity 117 member A
There are no transcripts associated with this gene.

Gene
SLC35B1
Gene Name
solute carrier family 35 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278784.1 Intron NP_001265713.1
NM_005827.2 Intron NP_005818.2
XM_005256929.3 Intron XP_005256986.1
XM_006721632.3 Intron XP_006721695.1
XM_011524179.2 Intron XP_011522481.1
XM_011524180.2 Intron XP_011522482.1

View Full Product Details