Product Details

SNP ID
rs141214654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1423671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGACCTGCAGGAGGAGAATAAGGA[C/G]AGCACTTTATTTCCAGGTAGGAATG
Phenotype
MIM: 164762
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRK PubMed Links

Gene Details

Gene
CRK
Gene Name
CRK proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005206.4 Intron NP_005197.3
NM_016823.3 Intron NP_058431.2

View Full Product Details