Product Details
- SNP ID
-
hCV25613705
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:1844502 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATATGTAAGGCATTTTTCTTTGCT[A/C]GCACAGGTATGAGTAGCTCTCAGAG
- Phenotype
-
MIM: 179835
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RPA1
PubMed Links
Gene Details
- Gene
- RPA1
- Gene Name
- replication protein A1
There are no transcripts associated with this gene.
View Full Product Details