Product Details

SNP ID
rs144267011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66932624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAGAGTCTTCTCAGGCCAGCTT[C/T]GGTCTAGGCTATAACCCATACTGAG
Phenotype
MIM: 605278 MIM: 614778 MIM: 179503
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CES2 PubMed Links

Gene Details

Gene
CES2
Gene Name
carboxylesterase 2
There are no transcripts associated with this gene.

Gene
FAM96B
Gene Name
family with sequence similarity 96 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016062.3 Intron NP_057146.1
Gene
RRAD
Gene Name
RRAD, Ras related glycolysis inhibitor and calcium channel regulator
There are no transcripts associated with this gene.

View Full Product Details