Product Details

SNP ID
rs114625980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19172565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGGCTACCAGCAGCCCTGAAA[A/G]TAGCAACACCTTGTTAGGATGTTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYT17 PubMed Links

Gene Details

Gene
SYT17
Gene Name
synaptotagmin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308157.1 934 UTR 5 NP_001295086.1
NM_016524.3 934 Intron NP_057608.2
XM_005255362.3 934 Intron XP_005255419.1
XM_011545865.1 934 Intron XP_011544167.1
XM_011545866.2 934 UTR 5 XP_011544168.1
XM_011545868.1 934 Intron XP_011544170.1
XM_011545869.2 934 Intron XP_011544171.1
XM_011545870.2 934 Intron XP_011544172.1
XM_011545871.2 934 Intron XP_011544173.1
XM_011545872.2 934 Intron XP_011544174.1
XM_017023280.1 934 Intron XP_016878769.1
XM_017023281.1 934 Intron XP_016878770.1

View Full Product Details