Product Details

SNP ID
rs35737774
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:147828042 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTCACCGATTTTCAAAACATAACAC[A/G]CATTCATTGGGATAAGTATTTCCAT
Phenotype
MIM: 167790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPINK1 PubMed Links

Gene Details

Gene
SPINK1
Gene Name
serine peptidase inhibitor, Kazal type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003122.4 463 Silent Mutation TGC,TGT C58C NP_003113.2

View Full Product Details