Product Details

SNP ID
rs41301331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153951801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCTGCCCTCACCTCCCTGGGTG[C/T]CCCCACCACCCCAGCACCAATTCGC
Phenotype
MIM: 300019 MIM: 312420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCFC1 PubMed Links

Gene Details

Gene
HCFC1
Gene Name
host cell factor C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005334.2 Intron NP_005325.2
XM_006724815.2 Intron XP_006724878.1
XM_006724816.2 Intron XP_006724879.1
XM_011531147.2 Intron XP_011529449.1
XM_011531148.2 Intron XP_011529450.1
XM_017029471.1 Intron XP_016884960.1
XM_017029472.1 Intron XP_016884961.1
Gene
RENBP
Gene Name
renin binding protein
There are no transcripts associated with this gene.

View Full Product Details