Product Details

SNP ID
rs58701628
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:22637572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGTGGGAGCTGGGTGTGAGTGT[A/G]ATGTCCAACCTGCCCAGGCCCTCCC
Phenotype
MIM: 120550 MIM: 120575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1QA PubMed Links

Gene Details

Gene
C1QA
Gene Name
complement C1q A chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015991.2 Intron NP_057075.1
Gene
C1QC
Gene Name
complement C1q C chain
There are no transcripts associated with this gene.

Gene
MIR6127
Gene Name
microRNA 6127
There are no transcripts associated with this gene.

View Full Product Details