Product Details
- SNP ID
-
rs12943620
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:80470858 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTCGAAGGTCCACTTGGTGGCCCC[G/T]CCGTAGATCTCGATGTGGGATTCAG
- Phenotype
-
MIM: 602367
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NPTX1
PubMed Links
Gene Details
- Gene
- NPTX1
- Gene Name
- neuronal pentraxin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002522.3 |
1412 |
Silent Mutation |
|
|
NP_002513.2 |
View Full Product Details