Product Details

SNP ID
rs36073794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157138869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTTGCTATCATCAGCATCCATT[A/G]GTTCAGTTTTTACTCTCATTCCTGC
Phenotype
MIM: 605045
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED7 PubMed Links

Gene Details

Gene
MED7
Gene Name
mediator complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100816.1 671 Missense Mutation CCA,CTA P188L NP_001094286.1
NM_004270.4 671 Missense Mutation CCA,CTA P188L NP_004261.1

View Full Product Details