Product Details

SNP ID
rs35799412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48720918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGTGCAAGGCATCGTCCGTCAC[G/T]GGACCAGTGAGGCGCAGGCGCGAGC
Phenotype
MIM: 610349 MIM: 609623
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAMSTR PubMed Links

Gene Details

Gene
MAMSTR
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
There are no transcripts associated with this gene.

Gene
RASIP1
Gene Name
Ras interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017805.2 2795 Silent Mutation NP_060275.2
XM_011527053.2 2795 Silent Mutation XP_011525355.2
XM_011527054.2 2795 Intron XP_011525356.2
XM_011527055.2 2795 Silent Mutation XP_011525357.2
XM_017026914.1 2795 Silent Mutation XP_016882403.1
XM_017026915.1 2795 Silent Mutation XP_016882404.1

View Full Product Details