Product Details

SNP ID
rs62000969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66590803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCTTGTCCCTCAAACCACTCT[A/C]GTCTCGCCAGCAATGCCTCCCGCAG
Phenotype
MIM: 603864
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC87 PubMed Links

Gene Details

Gene
CCDC87
Gene Name
coiled-coil domain containing 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018219.2 2281 Missense Mutation CGA,CTA R738L NP_060689.2
Gene
CCS
Gene Name
copper chaperone for superoxide dismutase
There are no transcripts associated with this gene.

View Full Product Details