Product Details

SNP ID
rs145176499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12807434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCACAGGGTCAAATACAACCT[G/T]AACTCCCTGTCACATGATACAGCCA
Phenotype
MIM: 600538 MIM: 606034
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRDX2 PubMed Links

Gene Details

Gene
PRDX2
Gene Name
peroxiredoxin 2
There are no transcripts associated with this gene.

Gene
RNASEH2A
Gene Name
ribonuclease H2 subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006397.2 584 Silent Mutation CTG,CTT L113L NP_006388.2
XM_006722619.2 584 Silent Mutation CTG,CTT L69L XP_006722682.1

View Full Product Details