Product Details

SNP ID
rs11554400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12806706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCGAGCTGGAGAGAGACAATAC[A/G]GGCCGCTGTCGCCTGAGTTCGCCTG
Phenotype
MIM: 600538 MIM: 606034
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRDX2 PubMed Links

Gene Details

Gene
PRDX2
Gene Name
peroxiredoxin 2
There are no transcripts associated with this gene.

Gene
RNASEH2A
Gene Name
ribonuclease H2 subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006397.2 93 Silent Mutation ACA,ACG T11T NP_006388.2
XM_006722619.2 93 Intron XP_006722682.1

View Full Product Details