Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001014443.2 | 507 | Missense Mutation | CCT,TCT | P91S | NP_001014443.1 |
NM_001319848.1 | 507 | Missense Mutation | CCT,TCT | P91S | NP_001306777.1 |
NM_012475.4 | 507 | Missense Mutation | CCT,TCT | P91S | NP_036607.3 |
XM_011509414.1 | 507 | Missense Mutation | CCT,TCT | P120S | XP_011507716.1 |
XM_017001036.1 | 507 | Missense Mutation | CCT,TCT | P120S | XP_016856525.1 |
XM_017001037.1 | 507 | Missense Mutation | CCT,TCT | P91S | XP_016856526.1 |
XM_017001038.1 | 507 | Missense Mutation | CCT,TCT | P91S | XP_016856527.1 |
XM_017001039.1 | 507 | Missense Mutation | CCT,TCT | P91S | XP_016856528.1 |
XM_017001040.1 | 507 | Missense Mutation | CCT,TCT | P120S | XP_016856529.1 |
XM_017001041.1 | 507 | UTR 5 | XP_016856530.1 | ||
XM_017001042.1 | 507 | Intron | XP_016856531.1 |