Product Details

SNP ID
rs11542317
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:23558989 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCAAAAGCTCCTTTTGTCGTTGG[A/C]GATGACAAGTTCCGGAGTGAGCTCG
Phenotype
MIM: 600277
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ID3 PubMed Links

Gene Details

Gene
ID3
Gene Name
inhibitor of DNA binding 3, HLH protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002167.4 699 Missense Mutation GCC,TCC A111S NP_002158.3

View Full Product Details