Product Details
- SNP ID
-
rs1038034466
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:206866714 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTAGCAACTAAACTCTCTTTCCTA[C/T]CTCCATTTAATGGATGGAGAAACTG
- Phenotype
-
MIM: 605619
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL20
PubMed Links
Gene Details
- Gene
- IL20
- Gene Name
- interleukin 20
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018724.3 |
|
Intron |
|
|
NP_061194.2 |
View Full Product Details