Product Details

SNP ID
rs11614892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52009252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAACAGCAGCGGTGAGTCACCAA[C/T]ACCCAGCCCCTGCCATGGTCCAAAG
Phenotype
MIM: 612027
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRASP PubMed Links

Gene Details

Gene
GRASP
Gene Name
general receptor for phosphoinositides 1 associated scaffold protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271856.1 7 Intron NP_001258785.1
NM_181711.3 7 Intron NP_859062.1
XM_005268691.3 7 Intron XP_005268748.1
XM_011537996.1 7 UTR 5 XP_011536298.1

View Full Product Details