Product Details

SNP ID
rs78998950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72249826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATTCCCCAAACCACCAAAAAAA[C/T]GCAGACTGAGATCAAATACACAACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP6 PubMed Links

Gene Details

Gene
PARP6
Gene Name
poly(ADP-ribose) polymerase family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323515.1 Intron NP_001310444.1
NM_001323516.1 Intron NP_001310445.1
NM_001323519.1 Intron NP_001310448.1
NM_001323521.1 Intron NP_001310450.1
NM_001323522.1 Intron NP_001310451.1
NM_001323523.1 Intron NP_001310452.1
NM_001323524.1 Intron NP_001310453.1
NM_001323525.1 Intron NP_001310454.1
NM_001323526.1 Intron NP_001310455.1
NM_001323528.1 Intron NP_001310457.1
NM_001323530.1 Intron NP_001310459.1
NM_001323531.1 Intron NP_001310460.1
NM_001323532.1 Intron NP_001310461.1
NM_020214.3 Intron NP_064599.2
XM_011521805.1 Intron XP_011520107.1
XM_017022418.1 Intron XP_016877907.1

View Full Product Details