Product Details

SNP ID
rs12051249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67207524 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGGTCTGCAGTAGAGGGAGGCAA[A/G]AAGCAACCCGGGTCAAGGCCAGGGT
Phenotype
MIM: 600659 MIM: 606422 MIM: 613701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
E2F4 PubMed Links

Gene Details

Gene
E2F4
Gene Name
E2F transcription factor 4
There are no transcripts associated with this gene.

Gene
ELMO3
Gene Name
engulfment and cell motility 3
There are no transcripts associated with this gene.

Gene
LOC105369155
Gene Name
uncharacterized LOC105369155
There are no transcripts associated with this gene.

Gene
LRRC29
Gene Name
leucine rich repeat containing 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004055.1 1133 Intron NP_001004055.1
NM_012163.2 1133 Intron NP_036295.1
XM_017023126.1 1133 UTR 3 XP_016878615.1
XM_017023127.1 1133 UTR 3 XP_016878616.1
XM_017023128.1 1133 UTR 3 XP_016878617.1
XM_017023129.1 1133 UTR 3 XP_016878618.1
XM_017023130.1 1133 UTR 3 XP_016878619.1
XM_017023131.1 1133 UTR 3 XP_016878620.1
XM_017023132.1 1133 UTR 3 XP_016878621.1
XM_017023133.1 1133 Intron XP_016878622.1
XM_017023134.1 1133 UTR 3 XP_016878623.1
Gene
MIR328
Gene Name
microRNA 328
There are no transcripts associated with this gene.

View Full Product Details