Product Details

SNP ID
hCV25761332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64366106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGAAGAGAGAACATCAATAGCC[A/C]TCCAAGTATCAACTGTTTGAAAAAA
Phenotype
MIM: 606274 MIM: 610696
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CSNK1G1 PubMed Links

Gene Details

Gene
CSNK1G1
Gene Name
casein kinase 1 gamma 1
There are no transcripts associated with this gene.

Gene
KIAA0101
Gene Name
KIAA0101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029989.2 Intron NP_001025160.1
NM_014736.5 Intron NP_055551.1

View Full Product Details