Product Details

SNP ID
rs74162890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:131972898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTTCTTGGTTTTTTTGTTTCGC[C/T]TTTTTGTTTTTTTTAAAGCAGGAAA
Phenotype
MIM: 603293 MIM: 613992
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BNIP3 PubMed Links

Gene Details

Gene
BNIP3
Gene Name
BCL2 interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004052.3 Intron NP_004043.3
Gene
PPP2R2D
Gene Name
protein phosphatase 2 regulatory subunit Bdelta
There are no transcripts associated with this gene.

View Full Product Details