Product Details

SNP ID
rs34804482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:27368192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAAATGTGAGTACATGCCACAG[C/T]CGTAGTAGAAATCCTGTTCCAGACA
Phenotype
MIM: 612324 MIM: 606666
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC34 PubMed Links

Gene Details

Gene
CCDC34
Gene Name
coiled-coil domain containing 34
There are no transcripts associated with this gene.

Gene
LGR4
Gene Name
leucine rich repeat containing G protein-coupled receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018490.2 3016 Missense Mutation GAC,GGC D844G NP_060960.2

View Full Product Details