Product Details

SNP ID
rs3745816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45609649 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAATTCCTGCCCTGACACCTGACT[C/T]TTCCCTGGCCGCATCAGACCACCCG
Phenotype
MIM: 600551
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EML2 PubMed Links

Gene Details

Gene
EML2
Gene Name
echinoderm microtubule associated protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193268.1 2721 UTR 3 NP_001180197.1
NM_001193269.1 2721 UTR 3 NP_001180198.1
NM_012155.2 2721 UTR 3 NP_036287.1
XM_006723125.2 2721 UTR 3 XP_006723188.1
XM_006723126.2 2721 UTR 3 XP_006723189.1
XM_011526694.2 2721 UTR 3 XP_011524996.1
XM_011526695.2 2721 UTR 3 XP_011524997.1
XM_011526698.2 2721 UTR 3 XP_011525000.1
XM_011526700.2 2721 UTR 3 XP_011525002.1
XM_011526701.2 2721 UTR 3 XP_011525003.1
XM_017026550.1 2721 UTR 3 XP_016882039.1
XM_017026551.1 2721 Intron XP_016882040.1
Gene
GPR4
Gene Name
G protein-coupled receptor 4
There are no transcripts associated with this gene.

View Full Product Details