Product Details

SNP ID
rs999565312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:77788937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGTCCAGTATCCGCGCCTTTCT[C/T]GCTTTCTCTCTTTTTGCGCGCTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VAT1L PubMed Links

Gene Details

Gene
VAT1L
Gene Name
vesicle amine transport 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020927.2 Intron NP_065978.1

View Full Product Details