Product Details

SNP ID
rs142617011
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:172420766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACTGGCCTGTTTACTTCGTCTC[C/T]TAACAAAAAACACTTTGGATTCAGG
Phenotype
MIM: 611445
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf105 PubMed Links

Gene Details

Gene
C1orf105
Gene Name
chromosome 1 open reading frame 105
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300760.1 2207 Intron NP_001287689.1
NM_139240.3 2207 UTR 5 NP_640333.3
XM_005245608.2 2207 Intron XP_005245665.1
XM_005245610.3 2207 Intron XP_005245667.1
XM_006711630.2 2207 Intron XP_006711693.1
XM_011510152.2 2207 UTR 5 XP_011508454.1
XM_011510153.2 2207 UTR 5 XP_011508455.1
XM_011510154.2 2207 UTR 5 XP_011508456.1
XM_011510155.2 2207 UTR 5 XP_011508457.1
XM_011510156.2 2207 UTR 5 XP_011508458.1
XM_017002808.1 2207 Intron XP_016858297.1
Gene
DNM3
Gene Name
dynamin 3
There are no transcripts associated with this gene.

View Full Product Details