Product Details
- SNP ID
-
rs3740312
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:75212023 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GAAACGAATTCCCCAAGTTTGTATA[G/A]TCATGGTTATCTGTTTTAGAAATAA
- Phenotype
-
MIM: 193245
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
VDAC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56115704] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- VDAC2
- Gene Name
- voltage dependent anion channel 2
View Full Product Details