Product Details

SNP ID
rs35696191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44557117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCTTTCCAGAAGCAAGAGGGG[C/T]GCCTGACTGTGAAGCAGGAGCCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZKSCAN7 PubMed Links

Gene Details

Gene
ZKSCAN7
Gene Name
zinc finger with KRAB and SCAN domains 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288590.1 495 Missense Mutation CGC,TGC R24C NP_001275519.1
NM_001288591.1 495 Missense Mutation CGC,TGC R24C NP_001275520.1
NM_001288592.1 495 Intron NP_001275521.1
NM_018651.3 495 Missense Mutation CGC,TGC R24C NP_061121.2
NM_025169.2 495 Missense Mutation CGC,TGC R24C NP_079445.1

View Full Product Details